Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Cold Mount Acrylmount Kit Acrylic Resin
SDP

Supplier:  Electron Microscopy Sciences 1256

Encompass_Preferred

500 g resin powder; 225 ml hardening liquid. Easy to use resins with short curing times and negligible shrinkage. Self-polymerizing components that harden with addition of catalyst. Hardened acrylic is thermoplastic/resistant to most chemicals.
Color: Dull yellowish, partly transparent
Curing time: 10 minutes
Hardness: 82 Shore D
Use: Routine examination
Shrinkage: 4

Catalog No. 50-980-303


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.