Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Acrylodan | 86636-92-2 | 95.1% | 225.29 g/mol | C15H15NO | 5 MG
SDP

Catalog No. 5000208528
Encompass_Preferred

Acrylodan is a thiol-reactive, environment-sensitive fluorescent dye that covalently labels cysteine residues and reports on local dipolarity and conformational dynamics within binding pockets such as Cys34.

  • Reacts with thiols to form a covalent fluorescent adduct.
  • Sensitive to local dipolarity and protein dynamics.
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic agitation may be required.
  • Purity approximately 95.1%; molecular weight 225.29 g/mol.
  • Store protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-08528 Supplier Medchemexpress LLC Supplier No. HYW0402305MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acrylodan is a thiol-reactive, environment-sensitive fluorescent dye that covalently labels cysteine residues and reports on local dipolarity and conformational dynamics within binding pockets such as Cys34.

  • Reacts with thiols to form a covalent fluorescent adduct.
  • Sensitive to local dipolarity and protein dynamics.
  • Soluble in DMSO at approximately 50 mg/mL; ultrasonic agitation may be required.
  • Purity approximately 95.1%; molecular weight 225.29 g/mol.
  • Store protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.