Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylonitrile, +99%, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
107-13-1
C3H3N
53.06
MFCD00001927
NLHHRLWOUZZQLW-UHFFFAOYSA-N
acrylonitrile,2-propenenitrile,vinyl cyanide,propenenitrile,cyanoethylene,carbacryl,fumigrain,acritet,acrylon,ventox
7855
CHEBI:28217
prop-2-enenitrile
C=CC#N
107-13-1
C3H3N
53.06
MFCD00001927
NLHHRLWOUZZQLW-UHFFFAOYSA-N
acrylonitrile,2-propenenitrile,vinyl cyanide,propenenitrile,cyanoethylene,carbacryl,fumigrain,acritet,acrylon,ventox
7855
CHEBI:28217
prop-2-enenitrile
C=CC#N
pH 5.9 to 7.6 (5% aq. soln.)
Infrared Spectrum Authentic
Refractive Index 1.3900 to 1.3920
Packaging Glass bottle
Quantity 1L
Boiling Point 77.3°C
Density 0.8000g/mL
Melting Point -83.5°C
Color Undesignated
Flash Point −0.2°C
Water 0.5% max.
Specific Gravity 0.8
Stabilizer 35 to 45ppm MEHQ
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula H2C=CHCN
Beilstein 02, 400
Fieser 01,14; 07,4; 13,317
Merck Index 15, 122
Solubility Information Solubility in water: 73.5g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Formula Weight 53.06
Physical Form Liquid
Percent Purity ≥99%
Chemical Name or Material Acrylonitrile
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
May cause cancer.
May cause respiratory irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger