Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrylonitrile, 99+%

Catalog No. AC149630100
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149630100 10 L Plastic drum
AC149631000 100 mL Glass bottle
AC149630010 1 L Glass bottle
AC149630025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AC149630100 Supplier Thermo Scientific Chemicals Supplier No. 149630100
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 107-13-1 | C3H3N | 53.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-13-1
Molecular Formula C3H3N
Molecular Weight (g/mol) 53.06
MDL Number MFCD00001927
InChI Key NLHHRLWOUZZQLW-UHFFFAOYSA-N
Synonym acrylonitrile, 2-propenenitrile, vinyl cyanide, propenenitrile, cyanoethylene, carbacryl, fumigrain, acritet, acrylon, ventox
PubChem CID 7855
ChEBI CHEBI:28217
IUPAC Name prop-2-enenitrile
SMILES C=CC#N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -83.5°C
Density 0.8000g/mL
pH 5.9 to 7.6 (5% aq. soln.)
Boiling Point 77.3°C
Flash Point −0.2°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Refractive Index 1.3900 to 1.3920
Linear Formula H2C=CHCN
Quantity 10 L
Beilstein 02, 400
Fieser 01,14; 07,4; 13,317
Specific Gravity 0.8
Merck Index 15, 122
Solubility Information Solubility in water: 73.5g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Stabilizer 35 to 45ppm MEHQ
Formula Weight 53.06
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Acrylonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
May cause cancer.
May cause respiratory irritation.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Toxic
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at
EINECSNumber 203-466-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.