Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester)

Catalog No. A20770
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20770 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20770 Supplier Invitrogen™ Supplier No. A20770
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester) is an amine-reactive molecule that enables crosslinking of acrylamide to proteins and other biomolecules containing amines.

Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester) is an amine-reactive molecule that enables crosslinking of acrylamide to proteins and other biomolecules containing amines. The succinimidyl ester of acryloyl-X covalently reacts with amines of proteins, amine-modified nucleic acids, and other biomolecules to yield acrylamides that can be copolymerized into polyacrylamide matrices or on surfaces, such as in microarrays and in biosensors. For instance, acryloyl-X, SE is used to prepare streptavidin acrylamide (Cat. No. S21379), which can copolymerize with acrylamide on a polymeric surface to create a uniform monolayer of immobilized streptavidin for binding to biotinylated ligands such as hybridization probes, enzymes, antibodies, and drugs.

Acryloyl-X, SE is a critical crosslinking reagent used in the super-resolution imaging techniques expansion microscopy (ExM) and protein-retention expansion microscopy (proExM) to add acrylamides to the amines of proteins of a preserved cell or tissue sample. This crosslinking with acrylamide allows the proteins to be anchored to a polymer hydrogel and polymerized. The polymer hydrogel is then swelled, allowing the cell or tissue sample to be physically expanded to improve spatial resolution in imaging. Expansion microscopy is a versatile super-resolution imaging technique that can be performed on many specimens, including 3D imaging of thick samples. This imaging technique also uses conventional microscopes, enabling easier and faster visualization of nanoscale biological structures without advanced super-resolution microscopes and equipment.

Properties of acryloyl-X, SE:

  • Synonyms – Acryloyl-X, SE; 6-((acryloyl)amino)hexanoic Acid, Succinimidyl Ester; 2,5-Dioxopyrrolidin-1-yl 6-Acrylamidohexanoate; N-Succinimidyl 6-Acrylamidohexanoate; N-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-2-propenamide
  • Molecular weight (MW) – 282.30
  • Molecular formula – C13H18N2O5
  • CAS number – 63392-86-9

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration of up to 10 mM.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Acryloyl-X SE
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Acryloyl-X
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.