Learn More
StatLab™ AcryMount Plus Mounting Media
Description
- Compatible with xylene and xylene-substitute clearing agents, including d-Limonene
- Slides mounted with Acrymount Plus may be screened in as little as 10 minutes
- Formulated to reduce oxidation, preventing yellowing and fading in H&E, IHC and special stains after coverslipping
- Maintains optimal refractive index for viewing and photo microscopy
- For labs seeking to eliminate final clearing with xylene, tissue may be coverslipped directly from absolute isopropyl alcohol
- Compatible with manual and most automated coverslipping processes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 16 oz. |
| Product Type | Mounting Medium |
| Packaging | Bottle |