Learn More
Electron Microscopy Sciences Acrytol Mounting Medium 16 OZ
SDP
Catalog No. 50365943
Encompass_Preferred
Acrytol is a rapid-drying mounting medium which enables slides to be screened immediately. It has a low viscosity, allowing the medium to flow easily. It also prevents air bubbles from becoming trapped. Acrytol contains an antioxidant to inhibit stain fading and prevent the formulation of annual rings. A thin coating of Acrytol is all that is needed to adhere a coverglass to a microscope slide.
Catalog No. 50-365-943 Supplier Electron Microscopy Sciences Supplier No. 13518
Acrytol is a rapid-drying mounting medium which enables slides to be screened immediately. It has a low viscosity, allowing the medium to flow easily. It also prevents air bubbles from becoming trapped. Acrytol contains an antioxidant to inhibit stain fading and prevent the formulation of annual rings. A thin coating of Acrytol is all that is needed to adhere a coverglass to a microscope slide.