Learn More
Aviva Systems Biology Corp ACSBG2 Antibody - middle region (ARP53793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSBG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACSBG2. Alternate names: BGR, BRGL, DKFZp434K1635, MGC111089, PRTD-NY3, PRTDNY3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-61130