Learn More
Aviva Systems Biology Corp ACSF2 Antibody - C-terminal region (ARP60794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human ACSF2. Alternate names: ACSMW, AVYV493, FLJ20920ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-165