Learn More
Aviva Systems Biology Corp ACSL1 Antibody - C-terminal region (ARP32784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ACSL1. Alternate names: ACS1, FACL1, FACL2, LACS, LACS1, LACS2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-54762