Learn More
Aviva Systems Biology Corp ACSL1 Antibody - N-terminal region (ARP32785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACSL1. Alternate names: ACS1, FACL1, FACL2, LACS, LACS1, LACS2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54763