Learn More
Cayman Chemical ACSL1humnrecombIntaa 20ug
SDP
Catalog No. 502943301
Encompass_Preferred
Source: Active recombinant human N-terminal His-tagged ACSL1 expressed in E. coli • Amino acids: 48-698 • MW: 74.62 kDa
Catalog No. 50-294-3301 Supplier Cayman Chemical Supplier No. 4041820 UG
Source: Active recombinant human N-terminal His-tagged ACSL1 expressed in E. coli • Amino acids: 48-698 • MW: 74.62 kDa