Learn More
Cayman Chemical ACSL3humnrecombIntaa 20ug
SDP
Catalog No. 502942209
Encompass_Preferred
Source: Active recombinant human N-Terminal His-tagged ACSL3 expressed in E. coli • Amino acids: 45-720 • MW: 77.19 kDa
Catalog No. 50-294-2209 Supplier Cayman Chemical Supplier No. 3999020 UG
Source: Active recombinant human N-Terminal His-tagged ACSL3 expressed in E. coli • Amino acids: 45-720 • MW: 77.19 kDa