Learn More
Aviva Systems Biology Corp ACSL5 Antibody - C-terminal region (ARP47232_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47232_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ACSL5. Alternate names: ACS2, ACS5, FACL5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59494