Learn More
Aviva Systems Biology Corp ACSM3 Antibody - N-terminal region (ARP60796_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60796_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSM3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ACSM3. Alternate names: SA, SAHApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-287-167