Learn More
ABclonal Technology ACSS1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15007500UL
Encompass_Preferred
This gene encodes a mitochondrial acetyl-CoA synthetase enzyme. A similar protein in mice plays an important role in the tricarboxylic acid cycle by catalyzing the conversion of acetate to acetyl CoA. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Catalog No. 50-001-04907