Learn More
Aviva Systems Biology Corp ACSS2 Antibody - N-terminal region (ARP61137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACSS2. It was validated on Western Blot. . Alternate names: ACAS2, ACECS, ACS, ACSA, DKFZp762G026, dJ1161H23.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-266