Learn More
TARGETMOL CHEMICALS INC ACSS2-IN-2 5MG
SDP
Catalog No. 502579685
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ACSS2-IN-2 (MTB-9655) is an inhibitor of acyl-CoA synthetase short-chain family member 2 (ACSS2). ACSS2-IN-2 can inhibit ACSS2 activity with an IC50 value of 3.8 nM. ACSS2-IN-2 can be used for the research of several diseases such as viral infection metabolic disorders neuropsychiatric diseases inflammatory/autoimmune conditions and cancer. purity: 99%
Catalog No. 50-257-9685 Supplier TARGETMOL CHEMICALS INC Supplier No. T600605MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ACSS2-IN-2 (MTB-9655) is an inhibitor of acyl-CoA synthetase short-chain family member 2 (ACSS2). ACSS2-IN-2 can inhibit ACSS2 activity with an IC50 value of 3.8 nM. ACSS2-IN-2 can be used for the research of several diseases such as viral infection metabolic disorders neuropsychiatric diseases inflammatory/autoimmune conditions and cancer. purity: 99%