Learn More
Medchemexpress LLC ACT-1004-1239 | 2178049-58-4 | 98.8% | C27H28F2N6O3 | 1 ML
SDP
Catalog No. 5000353571
Encompass_Preferred
ACT-1004-1239 is a potent, selective, orally active CXCR7 antagonist with an IC50 value of 3.2 nM. It is intended for research use only and not sold to patients.
  • Inhibits human, dog, rat, mouse, guinea pig, and macaque CXCR7 with IC50s ranging from 0.6 to 3.2 nM.
  • Promotes oligodendrocyte precursor cell (OPC) differentiation by increasing CXCL12 levels.
  • Increases plasma CXCL12 concentration in vivo.
  • Reduces disease severity in an EAE model.
  • Increases myelination in a Cuprizone-induced demyelination model.
Catalog No. 50-003-53571 Supplier Medchemexpress LLC Supplier No. HY14261710MM/1ML
ACT-1004-1239 is a potent, selective, orally active CXCR7 antagonist with an IC50 value of 3.2 nM. It is intended for research use only and not sold to patients.
  • Inhibits human, dog, rat, mouse, guinea pig, and macaque CXCR7 with IC50s ranging from 0.6 to 3.2 nM.
  • Promotes oligodendrocyte precursor cell (OPC) differentiation by increasing CXCL12 levels.
  • Increases plasma CXCL12 concentration in vivo.
  • Reduces disease severity in an EAE model.
  • Increases myelination in a Cuprizone-induced demyelination model.