Learn More
Medchemexpress LLC Act-1004-1239 | 2178049-58-4 | 98.8% | C27H28F2N6O3 | 10MG
SDP
Catalog No. 5000259673
Encompass_Preferred
ACT-1004-1239 is a potent, selective, orally available CXCR7 (ACKR3) antagonist developed as a first-in-class compound. It exhibits low nanomolar potency and has shown immunomodulatory and promyelinating effects in preclinical models and early clinical research.
  • Potent CXCR7 antagonism with IC50 3.2 nM.
  • Orally bioavailable compound suitable for systemic dosing.
  • Demonstrated immunomodulatory and promyelinating activity in vivo.
  • High chemical purity and solid, stable formulation for storage.
  • Compatible with in vitro and in vivo pharmacology studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-59673 Supplier Medchemexpress LLC Supplier No. HY14261710MG
ACT-1004-1239 is a potent, selective, orally available CXCR7 (ACKR3) antagonist developed as a first-in-class compound. It exhibits low nanomolar potency and has shown immunomodulatory and promyelinating effects in preclinical models and early clinical research.
  • Potent CXCR7 antagonism with IC50 3.2 nM.
  • Orally bioavailable compound suitable for systemic dosing.
  • Demonstrated immunomodulatory and promyelinating activity in vivo.
  • High chemical purity and solid, stable formulation for storage.
  • Compatible with in vitro and in vivo pharmacology studies.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).