Learn More
Medchemexpress LLC Act-389949 | 1258417-54-7 | 100.0% | 428.39 g/mol | C20H18F2N6O3 | 25MG
SDP
Catalog No. 5000239122
Encompass_Preferred
ACT-389949 is a potent, selective agonist of formyl peptide receptor type 2 (FPR2/ALX) used as a research reagent. Supplied as a powder for in vitro and in vivo studies, the compound has limited solution stability and is typically dissolved fresh; reported properties include an EC50 of about 3 nM for FPR2, molecular formula C20H18F2N6O3, and molecular weight 428.39 g/mol.
  • Potent, selective FPR2/ALX agonist with EC50 ≈ 3 nM.
  • High reported purity suitable for research applications.
  • Provided as a powder for stable storage at -20°C.
  • Soluble in DMSO for in vitro assays with reported protocols for in vivo formulations.
  • Suitable for pharmacological and mechanistic studies of FPR2 signaling.
Catalog No. 50-002-39122 Supplier Medchemexpress LLC Supplier No. HY12407125MG
ACT-389949 is a potent, selective agonist of formyl peptide receptor type 2 (FPR2/ALX) used as a research reagent. Supplied as a powder for in vitro and in vivo studies, the compound has limited solution stability and is typically dissolved fresh; reported properties include an EC50 of about 3 nM for FPR2, molecular formula C20H18F2N6O3, and molecular weight 428.39 g/mol.
  • Potent, selective FPR2/ALX agonist with EC50 ≈ 3 nM.
  • High reported purity suitable for research applications.
  • Provided as a powder for stable storage at -20°C.
  • Soluble in DMSO for in vitro assays with reported protocols for in vivo formulations.
  • Suitable for pharmacological and mechanistic studies of FPR2 signaling.