Learn More
Medchemexpress LLC ACT-660602 | 1646267-59-5 | C20H20F6N8OS | 1 ML
SDP
Catalog No. 5000375783
Encompass_Preferred
ACT-660602 is an orally active antagonist of chemokine receptor (CXCR3) with an IC50 value of 204 nM. It inhibits T-cell migration and shows efficacy in an acute lung injury model, making it suitable for autoimmune disease research.
  • Inhibits T-cell migration
  • Shows efficacy in an acute lung injury model
  • Selective for CXCR3 over hERG (IC50 of 18 μM for hERG)
  • Enhances metabolic stability
  • Displays non-competitive binding mode to CXCL10 and CXCL11
  • Exhibits anti-inflammatory activity
Catalog No. 50-003-75783 Supplier Medchemexpress LLC Supplier No. HY15109610MM/1ML
ACT-660602 is an orally active antagonist of chemokine receptor (CXCR3) with an IC50 value of 204 nM. It inhibits T-cell migration and shows efficacy in an acute lung injury model, making it suitable for autoimmune disease research.
  • Inhibits T-cell migration
  • Shows efficacy in an acute lung injury model
  • Selective for CXCR3 over hERG (IC50 of 18 μM for hERG)
  • Enhances metabolic stability
  • Displays non-competitive binding mode to CXCL10 and CXCL11
  • Exhibits anti-inflammatory activity