Learn More
Med Vet International ACT Drape, 55" x 77", Sterile, J1062D, Each
Numéro de catalogue. 5000583483
Encompass_Preferred
DuPont’s newer material, ACT (Advanced Composite Technology) is gaining popularity for drapes and gowns. It has an embossed pattern and is comparable to SMS material. The plain material can be autoclaved, but the impervious drape must be provided in sterilized pouches.
Numéro de catalogue. 50-005-83483 Fournisseur Med Vet International Code fournisseur JOR1062D
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
DuPont’s newer material, ACT (Advanced Composite Technology) is gaining popularity for drapes and gowns. It has an embossed pattern and is comparable to SMS material. The plain material can be autoclaved, but the impervious drape must be provided in sterilized pouches.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.