Learn More
Med Vet International ACT Drape, 77" x 104", Sterile, J1062F, Each
Catalog No. 5000583212
Encompass_Preferred
DuPont’s newer material, ACT (Advanced Composite Technology) is gaining popularity for drapes and gowns. It has an embossed pattern and is comparable to SMS material. The plain material can be autoclaved, but the impervious drape must be provided in sterilized pouches.
Catalog No. 50-005-83212 Supplier Med Vet International Supplier No. JOR1062F
DuPont’s newer material, ACT (Advanced Composite Technology) is gaining popularity for drapes and gowns. It has an embossed pattern and is comparable to SMS material. The plain material can be autoclaved, but the impervious drape must be provided in sterilized pouches.