Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ACT II Dual Culture Transport System

Catalog No. R124022 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R124022 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R124022 Supplier Thermo Scientific™ Supplier No. R124022

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.

Thermo Scientific™ Remel A.C.T. II Dual is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.

  • Fluids may be introduced through septum in cap
  • Sterile packaged in individual Tyvek™ envelope with two plastic-shaft rayon-tipped swabs for clean environments
Do not freeze or overheat. Do not incubate prior to use.
TRUSTED_SUSTAINABILITY

Specifications

Sterility Sterile
Swab Configuration Single Applicator
Description 2 plastic-shaft swabs; 16 x 125mm Tube; 10/Pk.
For Use With (Application) Healthcare
Quantity 10/Pk.
Shaft Material Plastic
Storage Requirements Store in original container at 15°C to 30°C until used.
Tip Material Rayon
Tube Size 16 x 125 mm

For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.