Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents MSM1445P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: HHF35. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Muscle or sarcoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: SDS extract of human myocardium. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: Multiple.Chromosome: 1q42.13 (ACTA1); 10q23.3 (ACTA2); 2p13.1 (ACTG2). Gene Symbol: ACTA1 (Skeletal); ACTA2 (Smooth); ACTG2 (Smooth). Gene ID: 70. Ensemble: ENSG00000159251. Uniprot: NP_005150.
Catalog No. 50-188-5414