Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 59MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: ACTA2/791. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Blood vessels in all tissues, smooth muscle or leiomyosarcoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human ACTA2 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 42kDa.Chromosome: 10q23.31. Gene Symbol: ACTA2. Gene ID: 59. Ensemble: ENSG00000107796. Uniprot: NP_001307784.
Catalog No. 50-188-5141