Learn More
AdipoGen Actagardin
SDP
Supplier:  AdipoGen AGCN20300M025
Encompass_Preferred
Chemical. CAS 59165-34-3. Formula C81H124N20O24S4. MW 1890.2. Isolated from Actinoplanes sp. Tetracyclic class II lantibiotic. Structurally related to mersacidine. Antibiotic. Antibacterial against Gram-positive bacteria and Gram-negative microorganisms. Specifically inhibits peptidoglycan synthesis in the cell wall of Gram-positive bacteria, resulting in the accumulation of both uridine 5-disphosphate-N-acetylmuramylpentapeptide and lipid II. Has the ability to form a complex with lipid intermediate II lipid II.
Catalog No. 50-256-2635