Learn More
Cayman Chemical ACTAmInophen GlucuronIdSo 10mg
SDP
Catalog No. 502941672
Encompass_Preferred
An inactive metabolite of acetaminophen; formed via glucuronidation of acetaminophen by UGT1A6, UGT1A9, UGT1A1, and UGT2B15
Catalog No. 50-294-1672 Supplier Cayman Chemical Supplier No. 2785010 MG
An inactive metabolite of acetaminophen; formed via glucuronidation of acetaminophen by UGT1A6, UGT1A9, UGT1A1, and UGT2B15