Learn More
Cayman Chemical Acetaminophen Glucuronide (sodium salt), Purity: ≥98%, CAS: 120595-80-4, 50 mg
SDP
Catalog No. 502940226
Encompass_Preferred
An inactive metabolite of acetaminophen; formed via glucuronidation of acetaminophen by UGT1A6, UGT1A9, UGT1A1, and UGT2B15
Catalog No. 50-294-0226 Supplier Cayman Chemical Supplier No. 2785050 MG
An inactive metabolite of acetaminophen; formed via glucuronidation of acetaminophen by UGT1A6, UGT1A9, UGT1A1, and UGT2B15