Learn More
Apexbio Technology LLC ACTB-1003 939805-30-8 50mg
SDP
Catalog No. 5000091140
Encompass_Preferred
ACTB-1003 (CAS 939805-30-8) also known as EOC317 is an orally bioavailable small-molecule kinase inhibitor that selectively targets fibroblast growth factor receptor 1 (FGFR1) vascular endothelial growth factor receptor 2 (VEGFR2) and the endothelial-specific tyrosine kinase Tie-2 In vitro studies demonstrate that ACTB-1003 suppresses the activity of these kinases at nanomolar concentrations By modulating these angiogenesis-associated signaling pathways ACTB-1003 serves as a valuable tool for investigating tumor angiogenesis and related signal transduction mechanisms in biomedical research
Catalog No. 50-000-91140 Supplier Apexbio Technology LLC Supplier No. B490450
ACTB-1003 (CAS 939805-30-8) also known as EOC317 is an orally bioavailable small-molecule kinase inhibitor that selectively targets fibroblast growth factor receptor 1 (FGFR1) vascular endothelial growth factor receptor 2 (VEGFR2) and the endothelial-specific tyrosine kinase Tie-2 In vitro studies demonstrate that ACTB-1003 suppresses the activity of these kinases at nanomolar concentrations By modulating these angiogenesis-associated signaling pathways ACTB-1003 serves as a valuable tool for investigating tumor angiogenesis and related signal transduction mechanisms in biomedical research