Learn More
Aviva Systems Biology Corp ACTB Antibody - middle region (ARP40174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human ACTB. Alternate names: BRWS1, PS1TP5BP1ApplicationIHC, WB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rat, Sheep, Zebrafish
Catalog No. 50-014-57318