Learn More
Medchemexpress LLC HY-P7453 10ug Medchemexpress, ACTB, His, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP745310UG
Encompass_Preferred
Medchemexpress, HY-P7453 10ug ACTB, His, Human CAS: Recombinant Human ACTB, His is produced by E. coli expression system. The target protein is expressed with sequence (Met1-Phe375) of Human ACTB fused with a His tag at the C-terminus. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3166