Learn More
Medchemexpress LLC HY-N6872 5mg Medchemexpress, Actein CAS:18642-44-9 Purity:>98%
SDP
Catalog No. 502082970
Encompass_Preferred
Medchemexpress, HY-N6872 5mg Actein CAS:18642-44-9 Actein is a triterpene glycoside isolated from the rhizomes of Cimicifuga foetida. Actein suppresses cell proliferation, induces autophagy and apoptosis through promoting ROS/JNK activation, and blunting AKT pathway in human bladder cancer. Actein has little toxicity in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2970 Supplier Medchemexpress LLC Supplier No. HYN68725MG
Medchemexpress, HY-N6872 5mg Actein CAS:18642-44-9 Actein is a triterpene glycoside isolated from the rhizomes of Cimicifuga foetida. Actein suppresses cell proliferation, induces autophagy and apoptosis through promoting ROS/JNK activation, and blunting AKT pathway in human bladder cancer. Actein has little toxicity in vivo. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.