Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ACTH (1-14) TFA | 96.05% | 1794.90 | 10 MG
SDP

Catalog No. 5000424337
Encompass_Preferred

ACTH (1-14) TFA is a fragment of adrenocorticotrophin, a tropic hormone produced by the anterior pituitary. It regulates cortisol and androgen production and is associated with conditions like Addison disease, Cushing syndrome, and Cushing disease.

  • Purity of 96.05%
  • Molecular weight of 1794.90
  • Chemical formula: C79H110F3N21O22S
  • Appears as a white to off-white solid
  • Sequence: Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly
  • Soluble in water at 100 mg/mL (55.71 mM)
  • Soluble in PBS at 25 mg/mL (13.93 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-24337 Supplier Medchemexpress LLC Supplier No. HYP1582A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACTH (1-14) TFA is a fragment of adrenocorticotrophin, a tropic hormone produced by the anterior pituitary. It regulates cortisol and androgen production and is associated with conditions like Addison disease, Cushing syndrome, and Cushing disease.

  • Purity of 96.05%
  • Molecular weight of 1794.90
  • Chemical formula: C79H110F3N21O22S
  • Appears as a white to off-white solid
  • Sequence: Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly
  • Soluble in water at 100 mg/mL (55.71 mM)
  • Soluble in PBS at 25 mg/mL (13.93 mM)
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.