Learn More
Medchemexpress LLC Acth (1-14) Tfa (Adrenocorticotropic Hormone Fragment 1-14 Tfa) | 25696-21-3 | 96.1% | 1794.90 | 5 MG
SDP
Catalog No. 5000422386
Encompass_Preferred
ACTH (1-14) TFA is a fragment of adrenocorticotropin, a tropic hormone produced by the anterior pituitary, which plays a crucial role in regulating cortisol and androgen production. This research-grade peptide is involved in various cellular functions and is associated with conditions like Addison disease, Cushing syndrome, and Cushing disease.
  • Regulates cortisol and androgen production
  • Fragment of adrenocorticotropin
  • Suitable for research applications
  • High purity for reliable experimental results
  • Available in various package sizes
Catalog No. 50-004-22386 Supplier Medchemexpress LLC Supplier No. HYP1582A5MG
ACTH (1-14) TFA is a fragment of adrenocorticotropin, a tropic hormone produced by the anterior pituitary, which plays a crucial role in regulating cortisol and androgen production. This research-grade peptide is involved in various cellular functions and is associated with conditions like Addison disease, Cushing syndrome, and Cushing disease.
  • Regulates cortisol and androgen production
  • Fragment of adrenocorticotropin
  • Suitable for research applications
  • High purity for reliable experimental results
  • Available in various package sizes