Learn More
Medchemexpress LLC HY-P1545A 1mg Medchemexpress, ACTH (1-17) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1545A1MG
Encompass_Preferred
Medchemexpress, HY-P1545A 1mg ACTH (1-17) (TFA) CAS: ACTH (1-17) TFA, an adrenocorticotropin analogue, is a potent human melanocortin 1 (MC1) receptor agonist with a Ki of 0.21 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0536