Learn More
Bachem ACTH (1-24) (human, bovine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4011958.0001
Encompass_Preferred
ACTH (1-24) (human, bovine, rat) Trifluoroacetate, H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH, 16960-16-0, C136H210N40O31S, Mr 2933.48, 1mg. Tetracosactide, Cosyntropin, Tetracosactrin. Tetracosactide, a C-terminally truncated ACTH, still exhibits corticotropic activity, but it is less immunogenic than the full-length peptide hormone. The peptide induces an increase in the circulating level of glucocorticoids.
Catalog No. 50-259-466