Learn More
Bachem ACTH (1-39) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4030325.1000
Encompass_Preferred
ACTH (1-39) (human) Trifluoroacetate, H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH, 12279-41-3, C207H308N56O58S, Mr 4541.13, 1mg. Corticotropin. The major regulator of adrenal cortex function. ACTH stimulates the synthesis of steroidal hormones.
Catalog No. 50-259-473