Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ACTH (11-24) acetate | 99.88% | 1652.04 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYP1558A10MG

Encompass_Preferred

ACTH (11-24) (acetate) is an adrenocorticotropic hormone (ACTH) receptor antagonist. It is a fragment of adrenocorticotropic and induces cortisol release. This compound can be used for research of the central nervous system. In vitro, ACTH (11-24) (acetate) elicits cortisol secretion submaximally in freshly dispersed or cultured beef adrenal cortical cells, and in vivo, it has slight influences on circulating plasma corticosterone values and on fighting behavior.

  • Adrenocorticotropic hormone (ACTH) receptor antagonist
  • Induces cortisol release
  • Suitable for research of the central nervous system
  • Purity of 99.88%
  • Molecular weight of 1652.04 (free base)

Catalog No. 50-004-22235


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.