Learn More
Medchemexpress LLC ACTH (11-24) acetate | 99.88% | 1652.04 (free base) | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP1558A5MG
Encompass_Preferred
ACTH (11-24) (acetate) is an adrenocorticotropic hormone (ACTH) receptor antagonist. It is a fragment of adrenocorticotropic hormone and induces cortisol release. This product can be used for research of the central nervous system.
  • Elicits cortisol secretion submaximally in freshly dispersed or cultured beef adrenal cortical cells.
  • Has slight influences on circulating plasma corticosterone values and on fighting behavior.
Catalog No. 50-004-22236