Learn More
Bachem ACTH (7-38) (human)
SDP
Supplier:  Bachem 4015541.1000
Encompass_Preferred
ACTH (7-38) (human), H-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-OH, 68563-24-6, C167H257N47O46, Mr 3659.17, 1mg. Corticotropin-Inhibiting Peptide (CIP).
Catalog No. 50-382-388