Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5443MSM9P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CLIP/1449. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1. Positive Control: Normal pituitary gland or pituitary tumor. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. Chromosome: 2p23.3. Gene Symbol: POMC. Gene ID: 5443. Ensemble: ENSG00000115138. Uniprot: NP_001306133.
Catalog No. 50-188-5070