Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 5443MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: 2F6. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Normal pituitary gland or pituitary tumor. Cellular Staining: Cytoplasm, Secreted. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Synthetic peptide corresponding to aa1-24 of human ACTH. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: ACTH is ~5kDa,, the POMC precursor is ~30kDa. The molecular weight of POMC depends upon isoform variation, post-translational modifications.Chromosome: 2p23.3. Gene Symbol: POMC. Gene ID: 5443. Ensemble: ENSG00000115138. Uniprot: NP_001306133.
Catalog No. 50-188-5065