Learn More
CPC Scientific H-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACTH002B
Encompass
ONE-LETTER SEQUENCE: RPVKVYPNGAEDESAEAFPLEFMOLECULAR FORMULA: C112H165N27O36MOLECULAR WEIGHT:2465.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [53917-42-3], RESEARCH AREA: HormonalREFERENCES: L.I. Larsson, Histochem., 55, 225 (1978)
Catalog No. 50-210-8595