Learn More
CPC Scientific H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACTH008A
Encompass
ONE-LETTER SEQUENCE: SYSMEHFRWGMOLECULAR FORMULA: C59H78N16O16S1MOLECULAR WEIGHT:1299.4STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [2791-05-1], RESEARCH AREA: Hormonal
Catalog No. 50-210-8606