Learn More
CPC Scientific H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACTH011B
Encompass
ONE-LETTER SEQUENCE: SYSMEHFRWGKPVGKKRMOLECULAR FORMULA: C95H145N29O23S1MOLECULAR WEIGHT:2093.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [7266-47-9], RESEARCH AREA: Hormonal
Catalog No. 50-210-8613