Learn More
CPC Scientific H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ACTH007B
Encompass
ONE-LETTER SEQUENCE: SYSMEHFRWGKPVMOLECULAR FORMULA: C75H106N20O19S1MOLECULAR WEIGHT:1623.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [22006-64-0], RESEARCH AREA: Hormonal
Catalog No. 50-210-8605