Learn More
CPC Scientific H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH 0.5MG
SDP
Supplier:  CPC Scientific ACTH001A
Encompass
Pituitary hormone N-terminal synthetic fragment that stimulates glucocorticoid and mineralocorticoid synthesis and release in the adrenal cortex.SEQUENCE: H-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OHONE-LETTER SEQUENCE: SYSMEHFRWGKPVGKKRRPVKVYPNGAEDESAEAFPLEFMOLECULAR FORMULA: C207H308N56O58S1MOLECULAR WEIGHT: 4541.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [12279-41-3]SYNONYMS: Adrenocorticotropic HormoneRESEARCH AREA: HormonalREFERENCES:D. Yamashiro et al, JACS, 95, 1310 (1973)N.Y. Ann. Acad. Sci., 297,1-641 (1977)SKU(s): ACTH-001A, ACTH-001B
Catalog No. 50-210-8592