Learn More
CPC Scientific H-Met-Glu-His-Phe-Arg-Trp-Gly-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ACTH009B
Encompass
ONE-LETTER SEQUENCE: MEHFRWGMOLECULAR FORMULA: C44H59N13O10S1MOLECULAR WEIGHT:962.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [4037-01-8], RESEARCH AREA: HormonalREFERENCES: R. Schwyzer et al., FEBS Letters, 19, 229 (1971)
Catalog No. 50-210-8609